VWR® Safety Scalpels
Supplier: VWR International
Certificates
About this item
Stainless steel, razor-sharp cutting edge for superior cutting and clean incisions.
- Wrapped in individual peel pouch
- Sterilized by gamma radiation
- Top activated retractable safety scalpel
Sterile. 10 per pack.
Certifications: ISO, GMP
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.