VWR® Scalpel Knife Round Handle
Supplier: VWR International
Certificates
About this item
These high-quality round scalpel knife handle fits the VWR® stainless and carbon steel scalpel blades.
- Round handle
- Secure blade fitment
- Autoclavable/sterilizable
- Ribbed/grooved
For classroom dissection, field-study, removal of small embedded objects and crafts.
Certifications: ISO, GMP
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.