VWR® Dissecting Delicate Scissors
Fournisseur: VWR International
Certificats
A propos de cet article
Fine, micro-point and sharp blades perfect for fine dissection and suture removal applications.
- Large rings for comfort, user control and stability
- Manufactured from AISI 420 stainless steel
- High polish, mirror finish
- Autoclavable and corrosion-resistant
Product conforms to international quality standards.
Certifications: ISO, GMP
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.