VWR® Premium Vannas Micro Dissecting Scissors
Fournisseur: VWR International
Certificats
A propos de cet article
These spring-action micro dissecting scissors are perfect for micro dissecting applications.
- Manufactured from ASTM-standard German stainless steel
- Anti-glare coating
- Autoclavable and corrosion-resistant
Top-tier instrument with superior craftsmanship. Product conforms to international quality standards.
Certifications: ISO, GMP
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.