PIPETMAX® Pipette Heads, Gilson
Fournisseur: GILSON, INC.
Certificats
A propos de cet article
PIPETMAX heads are specially designed for the PIPETMAX system.
The PIPETMAX® offers 8-channel pipette heads with the system. Choose from three options, pipetting volumes between 1 and 10 µl, pipetting volumes between 20 and 200 µl and fixed 1 ml volumes.
BodyTextAccessories: PIPETMAX D1200 Tip Reload Box and 4×1200 Tip Ejector pairs with the MAX4×1200 Pipette Head.
Customers Who Bought This Also Bought