Bond Elut™ 96 Square-Well Plates, 2 ml, Agilent Technologies
Fournisseur: AGILENT TECHNOLOGIES, INC (CSD) CA
Certificats
A propos de cet article
Bond Elut 96 square-well plates support well-plate SPE applications with larger sample volumes.
- Enhanced quality for trouble-free high throughput
- Recommended for 96-well plate applications with larger sample volumes
- Available with many of our most popular Bond Elut™ sorbents
- Added flexibility for method development
- Automation-friendly to free up operator time
- Compatible with the Agilent positive pressure manifold 96 processor (PPM-96)
Bond Elut™ 96 square-well plates facilitate transfer from cartridge-based methods to 96-well plate automated formats. The 2 ml wells accommodate the larger processing volumes from older methods, easing method transfer and revalidation. When larger samples or wash volumes are required, the 2 ml well volume offers flexibility.
Bond Elut™ 2 ml 96 square-well plates are designed to ensure compatibility with positive pressure and vacuum manifolds and is automation-friendly for trouble-free, high-throughput operation.
Customers Who Bought This Also Bought