NoTrax® 830 High-Voltage Switchboard Matting, Justrite®
Supplier: Justrite
Certificates
About this item
Corrugated Switchboard Matting is non-conductive, designed to provide insulation for the worker.
- Insulated matting engineered to protect workers from electric shock
- Electrically Insulative Type 2 PVC compound
- Class 2 Di-electric strength: 30000 volts
- Proof test of 20000 volts
- Corrugated top surface - Added traction and easy to sweep clean
Corrugated Switchboard Matting is a non-conductive floor matting made from a PVC compound, designed to provide insulation for the worker. The mat prevents the worker from being grounded, thereby eliminating the possibility of electrical shock. The corrugated saw-tooth top surface provides increased traction and is easy to sweep clean. Switchboard matting is intended for use in dry environments in front of high voltage boxes, electrical fuse boxes, and transformers.
Customers Who Bought This Also Bought