SP Bel-Art Filter Paper Discs for Buchner Table-Top Funnels, Bel-Art Products, a part of SP
Fournisseur: Bel-Art Products, a Part of SP
Certificats
A propos de cet article
These heavy bond grade filter paper discs are custom-sized to fit table-top Buchner funnels and have a thickness of 0.30 mm.
- Made from creped white cellulose to improve porosity
- Flow rate: 90 ml per minute
- Wet burst strength is greater than 68 KPa; able to retain particles as small as 3 microns
Customers Who Bought This Also Bought