ABS® Vaccine Refrigerators, Certified to the NSF/ANSI 456 Standard, Horizon Scientific
These vaccine refrigerators are certified to the NSF/ANSI 456 Standard for Vaccine Storage.
- Microprocessor temperature controller
- High / low temperature alarms
- Probe access port (³/₄")
- LED interior lighting
- Natural, hydrocarbon refrigerant
- Pharmacy toolkit and temperature logs
- Remote alarm contacts
- ENERGY STAR certified*
These units protect stored contents with greater stringency as outlined by the NSF/ANSI 456 Standard for Vaccine Storage. Units feature superior temperature stability and uniformity, quick recovery times after door openings, and forced air technology. These features help ensure that vaccines are kept safe and delivered at maximum effectiveness.
All units include a 2 year parts and labor warranty, plus an additional 3 year compressor parts warranty. They run on natural, hydrocarbon refrigerant for environmental health and economic efficiency. Ideal for pharmacy use.
Material: White powder coated steel
: UL, C-UL, ETL, C-ETL listed (either single or dual agency listings) and certified to UL471 standard, hydrocarbon refrigerant safety.
*Select models ENERGY STAR certified: 76516-540, 76516-544, 76516-538, and 76516-542
*Select models ENERGY STAR certified: 76516-540, 76516-544, 76516-538, and 76516-542
- Catalog No:
- 76516-544
- 76516-542
- 76516-540
- 76516-538
- Environmentally Preferable:
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Cat. No.:
- 76516-544
- 76516-542
- 76516-540
- 76516-538
- Defrost:
- Cycle optimized, zero energy defrost
- Cycle optimized, zero energy defrost
- Cycle optimized, zero energy defrost
- Cycle optimized, zero energy defrost
- Shelves/drawers:
- 5 shelves (four adjustable/one fixed) with guard rail on back
- 7 shelves (six adjustable/one fixed) with guard rail on back
- 5 shelves (four adjustable/one fixed) with guard rail on back
- 7 shelves (six adjustable/one fixed) with guard rail on back
- Door type:
- 1 swing solid door
- 1 swing solid door
- 1 swing glass door
- 1 swing glass door
- Electrical:
- 110 - 120 V, AC: 15 A (minimum)
- 110 - 120 V, AC: 15 A (minimum)
- 110 - 120 V, AC: 15 A (minimum)
- 110 - 120 V, AC: 15 A (minimum)
- Weight:
- 98 kg (216 lbs.)
- 98 kg (216 lbs.)
- 137 kg (302 lbs.)
- 137 kg (302 lbs.)
Frequently Bought Together