VWR® Auto Defrost Laboratory Freezers with Natural Refrigerants
Fournisseur: VWR International
Certificats
À propos de cet article
These auto defrost freezers are designd for multi-functional use in laboratory settings.
- Mechanical thermostat
- Adjustable shelving
- Probe access port
- Natural, hydrocarbon refrigerant
These VWR® General Purpose laboratory freezers feature natural, hydrocarbon refrigerant for environmental health and economic efficiency. All models come equipped with mechanical thermostats, adjustable shelving, probe access, and high density foam insulation.
All units include a two year parts and labor warranty, plus an additional three year compressor parts warranty.
Spécifications
- Catalog No:
- 76514-336
- 76514-334
- Environmentally Preferable:
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Cat. No.:
- 76514-336
- 76514-334
- Defrost:
- Auto
- Auto
- Electrical:
- 115 V, 60 Hz, 5 A, ¼ HP
- 115 V, 60 Hz, 5 A, ¼ HP
- Shelves/drawers:
- 4 adjustable shelves and 1 wire basket
- 3 adjustable shelves
- Door type:
- 1 solid swing door
- 1 solid swing door
- Material:
- Steel, powder coated
- Steel, powder coated
- Weight:
- 86 kg (190 lbs.)
- 82 kg (180 lbs.)