VWR® Standard Series Glass Door Laboratory Refrigerators
Fournisseur: VWR International
Certificats
A propos de cet article
These standard glass door laboratory refrigerators offer cold storage solutions at an economical pricepoint.
- Microprocessor temperature controller
- Digital temperature display
- Audible and visual temperature alarms
- Probe access port
- Keyed door lock
- Adjustable shelving
- Natural, hydrocarbon refrigerant
- ENERGY STAR certified*
These VWR® standard series refrigerators utilize microprocessor temperature controllers, digital temperature displays, and temperature alarms. The are designed for multi-functional use in laboratory settings. All models also come built with probe access, adjustable shelving, and interior lighting. Units utilize natural, hydrocarbon refrigerant for environmental health and economic efficiency.
All units include a two year parts and labor warranty, plus an additional three year compressor parts warranty.
Certifications: ETL listed.
*Select models ENERGY STAR certified: 76514-262, 76514-264, 76514-266, 76514-274
*Select models ENERGY STAR certified: 76514-262, 76514-264, 76514-266, 76514-274
Spécifications
- Catalog No:
- 76514-278
- 76514-260
- 76514-262
- 76514-270
- 76514-272
- 76514-274
- 76514-276
- 76514-264
- 76514-266
- 76514-268
- Environmentally Preferable:
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Cat. No.:
- 76514-278
- 76514-260
- 76514-262
- 76514-270
- 76514-272
- 76514-274
- 76514-276
- 76514-264
- 76514-266
- 76514-268
- Shelves/drawers:
- 12 adjustable shelves
- 3 adjustable shelves
- 3 adjustable shelves
- 8 adjustable shelves
- 8 adjustable shelves
- 8 adjustable shelves
- 12 adjustable shelves
- 4 adjustable shelves
- 4 adjustable shelves
- 8 adjustable shelves
- Material:
- Steel, powder coated
- Steel, powder coated
- Steel, powder coated
- Steel, powder coated
- Steel, powder coated
- Steel, powder coated
- Steel, powder coated
- Steel, powder coated
- Steel, powder coated
- Steel, powder coated
- Weight:
- 314 kg (693 lbs.)
- 99 kg (218 lbs.)
- 110 kg (243 lbs.)
- 161 kg (355 lbs.)
- 208 kg (458 lbs.)
- 221 kg (488 lbs.)
- 312 kg (688 lbs.)
- 134 kg (296 lbs.)
- 143 kg (315 lbs.)
- 167 kg (369 lbs.)