VWR® Plus Laboratory Freezers, Stainless Steel
Fournisseur: VWR International
Certificats
A propos de cet article
These VWR® Plus Series stainless steel freezers feature a multi-functional design for laboratory use.
- Microprocessor temperature controller
- Digital temperature display
- Audible and visual temperature alarms
- Probe access port
- Keyed door lock
- Forced draft air circulation
- Natural, hydrocarbon refrigerant
These VWR® freezers utilize state-of-the-art technology including microprocessor temperature controllers, digital temperature displays, and a series of temperature alarms. All models are equipped with probe access ports, adjustable shelving, and interior lighting. Units utilize natural, hydrocarbon refrigerant for environmental health and economic efficiency.
All units include a two year parts and labor warranty, plus an additional three year compressor parts warranty.
Certifications: ETL listed.
Spécifications
- Catalog No:
- 76514-214
- 76514-216
- 76514-218
- Environmentally Preferable:
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Cat. No.:
- 76514-214
- 76514-216
- 76514-218
- Defrost:
- Auto
- Auto
- Auto
- Electrical:
- 115 V, 60 Hz, 8 A, ¾ HP
- 115 V, 60 Hz, 8 A, ¾ HP
- 115 V, 60 Hz, 9 A, ¾ HP
- Shelves/drawers:
- 3 adjustable shelves
- 3 adjustable shelves
- 6 adjustable shelves
- Door type:
- 1 solid swing door
- 1 solid swing door
- 2 stainless steel swing doors
- Material:
- Stainless steel
- Stainless steel
- Stainless steel
- Weight:
- 134 kg (295 lbs.)
- 134 kg (295 lbs.)
- 204 kg (450 bs.)