VWR® Sterile Class 100 HS Nitrile Gloves
Fournisseur: VWR International
Certificats
A propos de cet article
The VWR® sterile Class 100 HS nitrile gloves are intended for use in cleanroom production and research environments. These hand-specific gloves are pair packed for easy donning.
- Gamma irradiated to SAL 10⁻⁶
- Made from premium compounded Nitrile
- Single use only
- Suitable for use in ISO Class 5 (FED-STD-209E Class 100) or higher cleanrooms
Manufactured from a synthetic copolymer of acrylonitrile, butadiene, and methylacrylic acid (components that comply with FDA CFR 21). High tensile strength and strong permeation resistance against a wide variety of solvents, acids, and bases.
Static-dissipative to inhibit harmful static discharge. Silicone-free material minimizes ionic contaminants and nonvolatile residue (NVR) transfer. AQL level of 1.5 for pinholes.
Packaging: Gloves are packaged 1 pair per pack, 10 packs per intermediate bag, 5 intermediate bags per master bag, 4 master bags per case. Packaging is made of 40% recycled material.
Spécifications
- Catalog No:
- 76458-126
- 76458-114
- 76458-116
- 76458-122
- 76458-124
- 76458-118
- 76458-120
- Ambidextrous/Hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Material:
- Nitrile
- Nitrile
- Nitrile
- Nitrile
- Nitrile
- Nitrile
- Nitrile
- Finish:
- Textured fingers
- Textured fingers
- Textured fingers
- Textured fingers
- Textured fingers
- Textured fingers
- Textured fingers
- Cuff style:
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Sterility:
- Sterile
- Sterile
- Sterile
- Sterile
- Sterile
- Sterile
- Sterile