EDVOTEK® Comprehensive Biotechnology LabStation™
# de catalogue 470336-114
Fournisseur: Edvotek
Spécifications
- Gradient:No
- Description:Comprehensive biotechnology LabStation™
- Cat. no.:470336-114
Spécifications
A propos de cet article
This extensive LabStation™ supports up to 48 students and offers a well-rounded introduction to the exciting field of biotechnology.
- Supports 48 students
The perfect equipment package to fit all your classroom biotech needs! LabStation™ includes all equipment needed for electrophoresis, PCR, ELISA, transformation etc.
Delivery: Includes 1× model 541-542 EdvoCycler™ 2 (48×0.2 ml); 6× model 502-504 M12 Complete™ Electrophoresis Package (14×7 cm Tray and 7 x 7 cm Trays (2)); 3× model 581 MV10 Protein Electrophoresis Apparatus; 3× model 5010-Q QuadraSource™ Power Supply (10 to 300 V for 1 or 4 units); 6× model 589 Variable Micropipette (0.5 to 10 µl); 6× model 591 Variable Micropipette (10 to 100 µl); 6× model 592-1 Variable Micropipette (100 to 1000 µl); 2× model 557 TruBlu™ LED Transilluminator; 10× model 969 Long Wave UV Mini-Light; 2 × model 539 3 L Digital Water Bath; 1× model 546 Incubation Oven; 3× model 534 Piccolo Microcentrifuge; 1× model 5023 Tornado Vortexer™; 3× model 635 Ultra Micropipet Tips (0.1 to 10 µl / 2 Racks of 96); 6× model 636 Yellow Micropipet Tips (1 to 200 µl / 2 Racks of 96); 3× model 637 Blue Micropipet Tips (200 to 1000 µl / 2 Racks of 100); 3× model 638 Fine Tip Micropipet Tips (1 to 200 µl / 1 Rack of 204).
Customers Who Bought This Also Bought