Borosil® Light Blocking Amber Glass Desiccators, Foxx Life Sciences
Fournisseur: Foxx Life Sciences
Certificats
A propos de cet article
From streamlined contours to interchangeable lids and bodies, Borosil® light blocking amber non-vacuum desiccators reflect great care and attention to detail.
- Material: Borosilicate glass 3.3
- Color: Amber
- Class/Quality grade: Type I
- With porcelain plate and borosilicate lid with plastic knob
These desiccators are perfect for drying, and for storage of moisture-sensitive substances. Compatible with any common desiccant. Available in inner diameters from 100 mm (XS) to 300 mm (XL).
Certifications: Complies with IS 6128.
Delivery: Supplied with a porcelain plate and ground glass flanges.
Customers Who Bought This Also Bought