ContactUsLinkComponent
CleanMax® Clean Manufactured Non Sterile Sleeves, Lakeland Industries
CleanMax® Clean Manufactured Non Sterile Sleeves, Lakeland Industries
# de catalogue 76492-452
Fournisseur:  Lakeland
Sleeves, tunneled elastic ends with thumb hole, Bound seams, Size: 18in length
CleanMax® Clean Manufactured Non Sterile Sleeves, Lakeland Industries
# de catalogue 76492-452
Fournisseur:  Lakeland
Spécifications
  • Length:
    18"
  • Color:
    White
  • Description:
    Clean manufactured sleeves with tunneled elastic ends
  • Cat. no.:
    76492-452
Spécifications
A propos de cet article
Clean manufactured nonsterile sleeves with bound seams. Clean manufactured in a silicone-free environment.
  • Fluid repellent; tested in accordance with ASTM D6978 for Chemotherapy Permeation
  • Open fold for easier donning
  • Resistant to blood, body fluid and viral penetration
  • Resistant to blood borne pathogens
  • IEST-RP-CC003 Category I Particle Cleanliness
  • Latex-free and silicone-free
CleanMax® cleanroom garments for controlled environments provide optimal comfort, quality, and protection. Ideally suited for use in ISO Class 4 - 8 cleanrooms and all controlled environments. CleanMax® garments are easy to don, comfortable to wear, and are backed by Lakeland's 30+ years as a manufacturer of disposable protective apparel.
Packaging: Packaged in pairs in a protective outer bag for ante areas.
Customers Who Bought This Also Bought