ESD Rubber Table Mats, Textured, Transforming Technologies
Supplier: Transforming Technologies
Certificates
About this item
The MT4000-Texture series ESD-safe 2-layer rubber matting is made of 100% co-extruded rubber.
- Textured surface
- Dissipative top layer, conductive bottom layer
- 0.080" thick
- Colors: Gray, green, blue, royal blue
The durable two layer mat is constructed with a textured dissipative top layer and a conductive bottom layer and offers excellent resistance to hot solders, soldering irons and most solvents. The MT-Texture series are available in 40' rolls in 24, 30, 36 and 48" widths. Pre cut mat sizes are available with grounding hardware and common point ground cords.
Certifications: Meets or exceeds requirements of ANSI ESD S541-2003. RoHS compliant.
Customers Who Bought This Also Bought