ESD Cleanroom Table Mats, Transforming Technologies
Fournisseur: Transforming Technologies
Certificats
A propos de cet article
Protect your sensitive items with this ultra-clean, particle-free, permanently static dissipative cleanroom ESD matting.
- Non-outgassing, non-blooming
- Contains no particulates, plasticizers or phthalates
- Permanently anti-static
The IDP-STAT 5500 series utilizes a patented inherently static dissipative polymer alloy system to provide clean, permanent ESD protection. The IDP-STAT 5500 series provides consistent static dissipation that is not dependent on humidity and will last the life of the mat. With minimal cleaning requirements, the IDP-STAT material is ideal for cleanroom settings. Engineered with superior durability and see-through clarity, the IDP-STAT 5500 is an ideal material for softwall panels to separate working areas in controlled environments such as cleanrooms and data centers.
Customers Who Bought This Also Bought