Dissipative V-Groove Vinyl ESD Floor Mats, Transforming Technologies
Fournisseur: Transforming Technologies
Certificats
A propos de cet article
Designed to provide a static-safe work area for a wide variety of applications where ESD properties are required.
- Dissipative ESD floor runner
- V-groove
- Gray
V-groove floor runner mats can reduce slips and falls by increasing traction, even when exposed to moisture and chemicals. Transforming Technologies’ VinylStat FM3 V-Groove solid floor runners are made from a homogeneous polymer mix of durable PVC.
Customers Who Bought This Also Bought