Conductive Rubber V-Groove Floor Mats, Black, Transforming Technologies
Supplier: Transforming Technologies
Certificates
About this item
This conductive ESD floor runner has a non-slip, grooved surface that provides excellent traction and allows the mat to be easily cleaned.
- Conductive ESD floor runner
- V-groove
- Gray
- ⅛" thick
The ribbed matting is also very resilient, allowing it to stand up to heavy foot traffic while still maintaining its ESD protection. The material also offers moderate chemical resistance. The matting is available in 4×32' rolls and an assortment of lengths. The Transforming Technologies GrooveSTAT™ Conductive Rubber Matting provides the electrical properties specified by ESDA standards for floor mats. Most common ESD runners available are made with dissipative vinyl, which does not comply with ESD standards.
Customers Who Bought This Also Bought