ESD Cleanroom Washable Face Masks, Transforming Technologies
Fournisseur: Transforming Technologies
Certificats
A propos de cet article
The FM03 is a reusable cleanroom safe, anti-static face mask.
- ESD fabric
- Cleanroom safe
- Washable
The mask is made with an ESD safe, Class 100 cleanroom fabric that features a conductive grid to reduces static generation. This face mask can be machine washed.
Customers Who Bought This Also Bought