ESD Rubber Table Mats, Smooth, Transforming Technologies
Fournisseur: Transforming Technologies
Certificats
A propos de cet article
Anti-static workstation mats are made of a durable 2-ply rubber that offers excellent resistance to hot solders.
- Dissipative work surface: 10E6 to 10E8 ohms Rtt
- Thickness 0.080"
- Resistance to ground 8×10E6 to 8 ohms
- Resistance point to point 2×10E6 to 8 ohms
Anti-Static workstation mats are made of a durable 2-ply rubber that offers excellent resistance to hot solders, soldering irons and most solvents.
A conductive layer backs the static dissipative top layer that creates outstanding electrical performance characteristics. Available in Nasa Blue, Royal Blue, Green or Gray with the following features:
Delivery: Includes two installed 10 mm ground snaps and a 15' low profile common point ground cord.
Customers Who Bought This Also Bought