Stainless Steel Cleanroom Waste Basket, Transforming Technologies
# de catalogue 76490-262
Fournisseur: Transforming Technologies
Spécifications
- Description:Conductive metal wastebasket
- Size:28 quart
- Cat. no.:76490-262
Spécifications
A propos de cet article
Conductive trash cans keep charged waste materials away from sensitive electronic devices and from chemicals that could be ignited by a spark.
- Ideal for cleanrooms and other controlled environments
- 7 gallon (28 quart) capacity
- Made with permanently conductive stainless steel
- Typical point-to-point resistance: 10² to 10³ ohm
When grounded, the cans and liners will not hold a static charge and will contain materials safely. This conductive ESD waste basket is a stylish rectangular wastebasket with radius corners and rolled edges. Stainless steel conductive waste baskets keep highly charged waste receptacles away from sensitive electronic devices. Ideal for cleanrooms, electronics manufacturing, inspection, quality control, and research laboratories. Very durable.
Customers Who Bought This Also Bought