Oxalate (C₂O₄)-2 Single-Element Ion Anion Standard, 1,000 µg/ml (1,000 ppm), VWR Chemicals BDH®
Supplier: VWR International
Certificates
About this item
Ion chromatography & ion selective electrode (ISE) standard.
- Single and multi-element standards for ion chromatography available
- Stable and traceable
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.