SteriPlast®/LaboPlast® Sampling Palette Knife Spatulas, Burkle
Fournisseur: BURKLE INC
Certificats
A propos de cet article
The sampling palette knife spatula can be used to apply, distribute, or evenly spread viscous media such as pastes and creams. The palette knife spatula reaches easily into the corners and curves of various containers and is suitable for scraping out bowls, removing residue or wiping off volumetric and dosing spoons.
- Made out of PS white, PS blue or Bio-PE
- Optionally packaged individually and sterilized
- For single use
- Clean room manufactured (clean room class 7)
The dimensionally stable long edge of the plastic scraper is ideal for cutting viscous media. A small tip on the long edge can be used for ripping, for example packaging film. The scraper is also suitable for stirring and blending.
The disposable sampling palette knife spatula is suitable for use in the pharmaceutical industry, chemistry, cosmetics, and food and beverage.
Certifications: Complies with EU food requirements and FDA regulations.
Customers Who Bought This Also Bought