Core Samplers, Burkle
Fournisseur: BURKLE INC
Certificats
A propos de cet article
The core sampler can be used to take samples of bulk materials, which can easily become stuck or settle during sampling due to their particle form.
- Stainless steel AISI 316 construction
- For bulk materials that become stuck or settle easily
- Order includes core sampler (consisting of boring rod and boring tube), fixing clip
In comparison to standard all-layer samplers for bulk materials, the two-part core sampler has no gap dimensions in which the medium can become stuck while it is being removed. This makes it possible to take a proper sample of powders and granulates through multiple layers, for example glass balls or metal powders.
Depending on the size, the stainless steel sampler can be used for a wide range of different containers, such as large bags or small containers. The bulk material sampler takes a representative core sample and is ideal for use in various laboratory and industrial applications.
Customers Who Bought This Also Bought