ContactUsLinkComponent
Sterilizing Tote Box, Natural, Polypropylene
Sterilizing Tote Box, Natural, Polypropylene
# de catalogue 470146-856
Fournisseur:  Dynalon
TRAY - 20X15X5 - 14.2L WLS82340-D
Sterilizing Tote Box, Natural, Polypropylene
# de catalogue 470146-856
Fournisseur:  Dynalon
Spécifications
  • Capacity:
    5 gal.s
  • Description:
    Sterilizing tote box
  • Material:
    PP
  • L×W×H:
    21.875×15.625×5.25"
  • Supplier No.:
    410555
Spécifications
A propos de cet article
Sterilizing tote boxes are molded from natural polypropylene, and are suitable for food use.
  • Chemically resistant to acids, alkalies, oils, and detergents
  • Nestable and stackable
  • Chip-resistant
  • Non-allergenic
  • Quick drying
  • Corrosion and mildew proof
  • Heavy-duty construction
Extra -tough injection molded polypropylene molded totes are designed for any place that needs a versatile and attractive storage system. Totes are heavy-duty, durable, and of the finest quality. Sterilizing tote boxes are molded of natural polypropylene and suitable for food use. These plastic tote boxes are chemically resistant to acids, alkalies, oils, and detergents and will not rust or corrode. Can be nested when stored.
Certifications: FDA, USDA approved.
Customers Who Bought This Also Bought