Air Cadet® Vacuum/Pressure Pumps, Antylia Scientific
Fournisseur: Argos Technologies
Certificats
A propos de cet article
Unique cavity configuration minimizes dead space and air entrapment with unique cavity configuration.
- No maintenance needed, AC motors are brushless with sealed ball bearings
- Worry-free pumping
All-plastic, convoluted diaphragm design handles many vacuum/pressure applications and reduces stress, wear, and heat build-up.
Spécifications
- Catalog No:
- 89492-594
- 89492-596
- 89492-598
- Noise:
- 72 dB(A); except model 07532-65: 76 dB(A)
- 72 dB(A); except model 07532-65: 76 dB(A)
- 72 dB(A); except model 07532-65: 76 dB(A)