ECHO Revolve 4, Hybrid Fluorescence Microscope (Upright/Inverted), Discover Echo
Supplier: DISCOVER ECHO INC.
Certificates
About this item
This 'all-in-one' hybrid microscope can be used for upright and inverted observation methods.
- 5 MP monochrome camera (upgrade from 3.2 MP) - this increases field of view and resolution for fluorescence imaging
- 5 MP CMOS color camera (brightfield)
- Intelligent objective nosepiece - this allows the software interface to automatically recognise the objective being used
- Illumination technique: Brightfield and Epi-fluorescence (phase contrast available as an option)
- Ultra-bright LEDs with 50000-hour lifespan
- Touch screen functions drive cameras and illumination, replacing manual turrets and levers
- Traditional eyepieces are replaced by an iPad® tablet for easier viewing (retina display)
- Custom software App simplifies the workflow of taking images and seamlessly integrates with other cloud sharing Apps
- Transmitted light, phase contrast, darkfield, polarized light and fluorescence in both upright and inverted configruations
The upright observation method is ideal for viewing glass slides. Useful for water dipping applications.
The inverted observation method is suitable for viewing live samples in fluidic chambers such as dishes and well plates. It also allows for easier manipulation of samples. Ideal for long working distance applications.
BodyTextAccessories: Z stack software module (optional) - automated acquisition of Z stacks.
Digital haze reduction software (optional).
Digital haze reduction software (optional).
Ordering information: To be completed with objectives, filters and light cubes, which are sold separately.
Delivery: Supplied with an iPad® interface, motorised epi-fluorescence turret, brightfield and fluorescence cameras, power supply, precision X/Y stage, microscope cover, dark box, stage clip and lens cloth.
Specifications
- Catalog No:
- 76376-718
- 76376-720
- 76376-726
- 76376-728
- 76376-722
- 76376-724
- 76299-602
- 76299-612
- 76299-610
- 76490-316
- 76490-302
- 76376-730
- 76376-732
- 76490-290
- 76490-310
- 76490-308
- Observation method:
- Polarised light, Phase contrast, Darkfield, Brightfield, Fluorescence
- Camera:
- 5 MP color (Brightfield), 5 MP monochrome camera (upgrade from 3.2 MP, fluorescence)
- Sensor:
- CMOS
- Display:
- 12.9" iPad® Retina® display, 2732×2048 (264 pixels/in.)
- Output:
- USB
- Electrical:
- 100 - 240 V (50/60 Hz)