ContactUsLinkComponent
BxPads Blue Sponges, Mortech®
BxPads Blue Sponges, Mortech®
# de catalogue 76498-608
Fournisseur:  Mortech Manufacturing
BxPADS Blue Sponges, 1000 Pack, BxPads are the thinnest biopsy sponges available to minimize pressure applied to unfixed specimens in biopsy cassettes.Dimensions:  1.25in X 1inQuantity:  1, 000 per bag
BxPads Blue Sponges, Mortech®
# de catalogue 76498-608
Fournisseur:  Mortech Manufacturing
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Dimensions:
    1.25×1"
  • Description:
    BxPads blue sponges
  • Cat. no.:
    76498-608
Spécifications
A propos de cet article
BxPads are the thinnest biopsy sponges available.
Designed to minimize pressure applied to unfixed specimens in biopsy cassettes.
Packaging: 1000 pads per bag.
Customers Who Bought This Also Bought