ContactUsLinkComponent
Vacuum Hydro Aspirator, Mortech®
Catalogue # 76498-430
Fournisseur:  Mopec
undefined
Vacuum Hydro Aspirator, Mortech®
Catalogue # 76498-430
Fournisseur:  Mopec
Supplier Number:  BE013
Spécifications
  • Description:
    Vacuum hydro aspirator
  • Cat. no.:
    76498-430
Spécifications
À propos de cet article
Deluxe hydro-aspirator with built-in vacuum breaker and control lever to be used to reverse the flow of water so as to direct it through the hose connections for either drawing fluid from the body cavity or rinsing a desired location.
  • Fits most standard faucet threads
  • Fabricated from chrome-plated brass
Unit can be purchased individually, or part of a whole water control unit.