ContactUsLinkComponent
Amputation Dissecting Board, Mortech®
Amputation Dissecting Board, Mortech®
# de catalogue 76498-304
Fournisseur:  Mortech Manufacturing
Amputation Dissecting Board, offers safe and secure dissection of any amputated limb specimen, securely sits on any table surface and heavy gauge stainless steel clamps hold any amputation specimen in place. Dimensions:  11in L (27 cm) x 6in W, Material:  PE with SS clamps, Color:  Opaque white
Amputation Dissecting Board, Mortech®
# de catalogue 76498-304
Fournisseur:  Mortech Manufacturing
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Amputation dissecting board
  • L×W:
    11×6"
  • Cat. no.:
    76498-304
Spécifications
A propos de cet article
Designed by Pathologist David Pinkhasov, this uniquely designed product offers safe and secure dissection of any amputated limb specimen.
  • 15" (38 cm) height
  • Polyethylene construction with stainless steel clamps
  • Opaque white finish
The Amputation dissection board securely sits on any table surface and features heavy gauge stainless steel clamps to hold any amputation specimen in place.
Customers Who Bought This Also Bought