Cork Dissecting Board with Optional Feet, Mortech®
Fournisseur: Mortech Manufacturing
Certificats
A propos de cet article
Neoprene cork surface offers durability during dissection and is fluid-resistant.
- 0.25" layer of neoprene cork permanently cemented to a 0.75" marine plywood base
- Durable and fluid-resistant cork construction
Removable non-skid legs provide an ergonomic, multi-user work surface height.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.