Borosil® Weighing Bottles with Interchangeable Ground Glass Stopper, Foxx Life Sciences®
Fournisseur: Foxx Life Sciences
Certificats
A propos de cet article
Borosil® Laboratory bottles are chemically resistant and stable. Fitted with interchangable glass stopper and are autoclavable and sterilizable.
- Borosilicate glass 3.3
- Mechanically strong and chemical resistant
- Round
- Glass stopper
Certifications: IS 1574.
Customers Who Bought This Also Bought