Borosil® Borosilicate 3.3 Low Expansion Glass, Round Bottom Boiling Flask
Fournisseur: Foxx Life Sciences
Certificats
A propos de cet article
Borosil® round bottom boiling flask.
- Matt-finished enameled area for marking
- Mechanically strong and chemical resistant
- Uniform wall thicknes, ideal for heating
Borosil® Flasks are designed to deliver the best combination of thermal shock resistance, physical strength and resistance to chemical attack. They are made so as to withstand the demands of the modern laboratory.
Certifications: Complies with ISO 1773.
Customers Who Bought This Also Bought