Body Elevation Support Slats, Mortech®
Body elevation support slats
Catalog # 76498-302
Supplier: Mopec
Fournisseur #: BC011
Jump to:
Product Details & Documents
These four body slats can be used to elevate the body on a table during any autopsy or mortuary preparation.
- Dimensions: 22.625×2.875" (55×5 cm)
- Chemically resistant polyethylene
- Four slats per pack
Elevation slats provide unobstructed drainage of fluids. They can be used in any anatomical location. Easily cleaned with Mopec Bench and Surface Cleaner. The suggested placement of the slats are head, shoulders, buttocks, and heels.
Spécifications
- Désignation:Body elevation support slats