ContactUsLinkComponent
Stainless Steel Dissecting Tray, Mortech®
Stainless Steel Dissecting Tray, Mortech®
# de catalogue 76498-300
Fournisseur:  Mortech Manufacturing
Dissecting Drain Tray, Stainless Steel, multi-purpose unit, used to confine spills/contamination to restricted area, standard dissecting boards will fit into 1in deep tray, sloped end overhangs any sink, allowing all fluids to drain directly into sink, Dimensions: 25in Long x 1in Wd x 1in Deep
Stainless Steel Dissecting Tray, Mortech®
# de catalogue 76498-300
Fournisseur:  Mortech Manufacturing
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Stainless steel dissecting tray
  • Cat. no.:
    76498-300
  • L×W×D:
    25×18×1"
Spécifications
A propos de cet article
A multi-purpose unit, this durable tray can be used to confine spills or contamination to a restricted area.
  • All stainless steel
  • Dissecting tray with sloped angle end for draining
All of Mopec’s standard dissecting boards will fit into this 1" deep tray. The sloped end overhangs any sink, allowing all fluids to drain directly into the sink.
Customers Who Bought This Also Bought