ContactUsLinkComponent
Stainless Steel Headrest, Mortech®
Stainless Steel Headrest, Mortech®
# de catalogue 76498-240
Fournisseur:  Mortech Manufacturing
Headrest, Stainless Steel, more natural cranial positioning during autopsy and mortuary procedures, broad convex curve fits within the arch of the neck, allowing the cadaver head to tilt up and rest upon work surface. Dimensions:  13.75in LongMaterial:  ?SS4 non-slip foot pads for stability
Stainless Steel Headrest, Mortech®
# de catalogue 76498-240
Fournisseur:  Mortech Manufacturing
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Stainless steel headrest
  • Cat. no.:
    76498-240
Spécifications
A propos de cet article
Designed to provide a more natural cranial positioning during autopsy and mortuary procedures.
  • 13.75" length
  • Stainless steel construction
  • Four non-slip foot pads for stability
The broad convex curve fits within the arch of the neck, allowing the cadaver head to tilt up and rest upon a work surface. This configuration allows easy access to the neck and ears without interference.
Customers Who Bought This Also Bought