ContactUsLinkComponent
JC-1 Mitochondrial Membrane Potential Detection Kit, Biotium
JC-1 Mitochondrial Membrane Potential Detection Kit, Biotium
# de catalogue 89138-884
Fournisseur:  Biotium
Numéro CAS:  
JC-1 Mitochondrial Detect. Kit
JC-1 Mitochondrial Membrane Potential Detection Kit, Biotium
# de catalogue 89138-884
Fournisseur:  Biotium
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    JC-1 Mitochondrial Membrane Potential Detection Kit
  • Cat. no.:
    89138-884
  • No. of Reactions:
    100
  • Supplier No.:
    30001
Spécifications
A propos de cet article
JC-1 dye is used for fluorescence-based measurement of mitochondrial membrane potential changes in cells, a key measure of cell viability.
  • Rapid, simple measurement of mitochondrial membrane potential/cell viability
  • Healthy cells stain red & green, dead cells stain green
  • For fluorescence microscopy, flow cytometry, or fluorescence microplate reader
  • Colors Green/Red
JC-1 Mitochondrial Membrane Potential Detection Kit is used to measure mitochondrial membrane potential changes in cells, a key indicator of cell viability.
The loss of mitochondrial membrane potential is a hallmark for apoptosis. It is an early event preceding phosphatidylserine externalization and coinciding with caspase activation. In non-apoptotic cells, JC-1 accumulates as aggregates in the mitochondrial membranes, resulting in red fluorescence. The brightness of red fluorescence is proportional to the potential and varies among different cell types. However, in apoptotic and necrotic cells, which have diminished mitochondrial membrane potential, JC-1 exists in the green fluorescent monomeric form. This kit provides a step-by-step protocol and ready-to-use reagents for performing assays for flow cytometry, fluorescence microscopy, or fluorescence microplate reader.
Spectral properties:
Ex/Em (cytoplasm): 510/527 nm (green)
Ex/Em (polarized mitochondria): 585/590 nm (red)
Kit components:
JC-1, 100X in DMSO
10X Assay Buffer
Caution: Store at 4 °C
Customers Who Bought This Also Bought