VWR® Sterile Masks
Fournisseur: VWR International
Certificats
A propos de cet article
VWR® made in USA sterile masks are an ideal option for single-use needs.
- Individually bagged
- Available with earloop or tie configurations
- 3-ply construction
- Gamma irradiated to 10⁻⁶
- Enclosed plastic encapsulated nose wire
A key part of the VWR® made in USA PPE line, these sterile cleanroom masks provide a comfortable fit while meeting sterility and filtration requirements of many controlled environments.
Packaging: Each mask individually bagged sealed. 10 bags per poly pack. 20 packs per case. Case is poly lined.
Customers Who Bought This Also Bought