CE Duo® CleanTech® Sterile 6-Pack with Bucket Liner, Vileda Professional®
# de catalogue 76499-312
Fournisseur: Vileda Professional - FHP
Spécifications
- Description:Sterile CE Duo® CleanTech® mop; single-use, double-sided microfiber cleanroom foam mop
- Cat. no.:76499-312
- Packaging:6/pack, 8 packs/case
- Length:19.7"
Spécifications
A propos de cet article
Mop is available in a conveinent six pack with a bucket liner, cutting down not only on the time it takes to prepare the mops but also significantly reducing the amount of packaging waste.
- Microfiber, overstitched cleanroom foam for cleaning/disinfecting floors, walls, and ceilings
- Adhesive free lamination techniqies are used to prevent additional contamination
- Excellent microfiber cleaning results have been independently proven
- Validated to 10⁻⁶ SAL per ISO 11137
- Reduce packaging waste
- Reduce amount of time needed to pre-prepare the mops
The Sterile CE Duo® CleanTech® mop is a single-use, double-sided microfiber cleanroom foam mop for cleaning and disinfecting floors, walls and ceilings. Now available in a conveinent 6 pack with included bucket liner.
Packaging: Packaged 6 sterile mops per pack, double bagged with bucket liner.
Customers Who Bought This Also Bought