CE Compact Trolley, Vileda Professional®
Catalog # 76499-310
Supplier: Vileda Professional - FHP
Specifications
- Description:Compact, modular, and stowable trolley; designed for use with pre-prepared mops and cleaning supplies
- Cat. no.:76499-310
Specifications
About this item
The CE Compact Trolley has been designed for use in controlled environments where footprints and becoming smaller or where storage is a concern.
- Electropolished stainless steel for a contamination free surface
- Autoclavable
- Low profile design fits under work stations or tables
- Fits full sized pre-prep boxes and buckets
- Reduces the risk of cross contamination by staying inside the controlled space for storage
- Accessories can be color coded
Compact, modular, and stowable trolley designed for use with pre-prepared mops and cleaning supplies, that stays inside the space and is adjustable to a variety of uses.
Customers Who Bought This Also Bought