Rotational Viscometers, ROTAVISC HELI Complete Series, IKA
Fournisseur: IKA Works
Certificats
A propos de cet article
The ROTAVISC HELI series determines the viscosity of liquids in all areas of application ranging from the laboratory to quality control. The four devices measure in different viscosity ranges.
- Large TFT display allows for an intuitive menu guidance
- Digital spirit level supports the correct set-up of the start-up condition
- Ramp function simplifies repetitive tasks
- Very high measurement accuracy
- Stepless speed adjustment
- Timer and graph function
- HELISTAND moves the mounted viscometer and its rotating body slowly and uniformly up and down in the sample
The level of measurement accuracy of the ROTAVISC HELI rotational viscometer for both Newtonian and non-Newtonian fluids is ±1% of the measuring range. The reproducibility is ±0,2%.
Ordering information: ROTAVISC HELI complete packages include SP 1 - 4 (lo-vi)/SP 7 - 12 (me-vi, hi-vi I, hi-vi II), protective bracket, temperature sensor, quick connector, hook connector and HELISTAND stand including a T-spindle set.
Spécifications
- Catalog No:
- 76499-014
- 76499-012
- 76499-018
- 76499-016
- Model:
- ROTAVISC Me-vi
- ROTAVISC Lo-vi
- ROTAVISC Hi-vi II
- ROTAVISC Hi-vi I
- Accuracy:
- 1%
- 1%
- 1%
- 1%
- Display:
- TFT, (4.3")
- TFT, (4.3")
- TFT, (4.3")
- TFT, (4.3")
- Interfaces:
- USB and RS232 Interface
- USB and RS232 Interface
- USB and RS232 Interface
- USB and RS232 Interface
- Electrical:
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- Operating temperature:
- −100...300 °C
- −100...300 °C
- −100...300 °C
- −100...300 °C
- W×D×H:
- 351×372×629 mm
- 351×372×629 mm
- 351×372×629 mm
- 351×372×629 mm