Surety Columns, Caplugs
Fournisseur: Caplugs
Certificats
A propos de cet article
Surety columns are ideal for use in removing fibrin and macro particles from serum to prevent line clogging.
- Surety columns have integral caps that fit directly onto sample cups
Choose from sizes ranging in size from 2.4 to 5.0 ml capacity.
Customers Who Bought This Also Bought