Specifications
- Color:Transparent
- Description:Honeycomb grid ESD curtain
- Size:4×98'
- Thickness:0.3 mm
- Cat. no.:76474-658
Specifications
About this item
Anti-Static and ESD curtains are a great way to separate critical workspaces while also attending to the static-sensitive nature of certain products.
- Ideal barrier wall for sensitive environments
- Honeycomb conductive scrim grounds static charges
- Curtain rolls can be easily converted for specific applications
- Easily cut custom sizes with a razor or knife and replace as needed
- Made with clear PVC
Transparent PVC ESD curtains are an ideal barrier wall for sensitive environments. Honeycomb grids are installed on both sides of the PVC curtain for safe dissipation of static electricity. These anti-static curtains are critical for applications where any electrical discharge could pose damage to critical components or explosion risk such as electronics, pharmaceutical, medical, printing, or painting. The ESD curtains also have a great draping ability so that they can be used as a protective covering for carts, instruments, or entire walls. The PVC material is very flexible and easy to cut for specific applications. Sold in 4' wide rolls.