Wolfgang Warmbier WT5000 Walking Test Kit, Transforming Technologies
# de catalogue 76474-634
Fournisseur: Transforming Technologies
Spécifications
- Description:WT5000 Walking test kit with computer interface
- Cat. no.:76474-634
Spécifications
A propos de cet article
A complete system to measure the body voltage generated while walking.
- Test and graph the performance of ESD floors in combination with a person
- Operates per ANSI/ESD STM 97.2. and IEC Standard 61340-4-5
The WT5000 Walking Test Kit has been designed to perform a voltage measurement of floor materials and footwear in combination with a person according to ANSI/ESD STM 97.2. and IEC Standard 61340-4-5. This test will evaluate the performance of the floor/footwear system. This test is of special importance when the floor/footwear system is used as a primary means for grounding personnel.
Customers Who Bought This Also Bought