A propos de cet article
A convenient hand-held device designed to locate and measure electrostatic fields on surfaces.
- Measures static voltages within ±20 kV (20000 V)
- Two LED guide ring lights help position the field meter
- Storage bag including ground cable and clip
- Pocket-size, portable design
- Battery operated
The EFM51 is a full-featured, hand-held field meter used to perform static surveys and evaluate charge accumulation in electronic manufacturing and industrial operations. The EFM51 measures static voltages within ±20 kV (20000 V) with a microcontroller that calculates the field strength (V/m) with a pre-selected distance (1, 5, 10 and 20cm). Results are displayed via a 12-digit alphanumeric LCD screen. Two LED guide ring lights help position the field meter at the right distance from a charged test object.
Unit is also available with charge plate, which is charged manually via the handheld high voltage generator.
The Walking Test Kit provides measurement data on the actual charge on a person while walking which is used to determine the effectiveness of the floor/footwear grounding system. The kit includes the EFM51 static field meter, walking test adapter plate, hand probe, and conductive carrying case.