VinylStat E Three-Layer ESD Table Mats with Foam Back, Transforming Technologies
Fournisseur: Transforming Technologies
Certificats
A propos de cet article
Vinyl ESD work surface matting with a foam back that provides extra cushion.
- Provides extra cushion for component safety and operator comfort
- Three layers of vinyl with excellent ESD protection
- Surface resistivity is in the dissipative range: 10⁷ to 10⁸
Designed for use at workstations and other manufacturing areas, the three-layer VME Vinyl serves a dual purpose: the closed-cell foam absorbs and reduces the shock and impacts associated with manufacturing electronic equipment such as disk drives and hard drives, while also dissipating static through immediate touch prior to handling such static-sensitive equipment. Made from two layers of polyvinyl chloride foam that encase a layer of conductive fleece to form an expanded static dissipative work surface.
Resistant to degradation by inorganic acids, organic acids, reducing agents, detergent solutions, alcohols, aliphatic hydrocarbons, mineral oil, and aldehydes.
Certifications: Meets or exceeds requirements of ANSI/ESD S20.20 per ANSI/ESD STM4.1.
Customers Who Bought This Also Bought