VinylStat C Homogeneous Vinyl ESD Table Mats, Transforming Technologies
Fournisseur: Transforming Technologies
Certificats
A propos de cet article
Single-layer vinyl homogeneous ESD work surface mat is ideal for general use.
- Surface resistivity is in the dissipative range: 10⁷ to 10⁸
- Homogeneous polymer mix construction
- Available in blue or grey; supplied as mat rolls
A homogeneous polymer mix construction with unsurpassed electrical properties gives VinylStat C a consistent, controlled static drain and physical properties offering the highest level of protection. Available in blue or grey.
Resistant to degradation by inorganic acids, organic acids, reducing agents, detergent solutions, alcohols, aliphatic hydrocarbons, mineral oil, amines, and aldehydes.
Customers Who Bought This Also Bought