ContactUsLinkComponent
ViaVac™ Red/Green (FUN®1), Biotium
ViaVac™ Red/Green (FUN®1), Biotium
# de catalogue 75832-630
Fournisseur:  Biotium
Numéro CAS:  
ViaVac Red/Green, 100ul
ViaVac™ Red/Green (FUN®1), Biotium
# de catalogue 75832-630
Fournisseur:  Biotium
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    ViaVac™ red/green (FUN®1)
  • Size:
    100 µl
  • Cat. no.:
    75832-630
  • Supplier No.:
    29068
Spécifications
A propos de cet article
A fluorescent vitality dye for yeast, identical in structure to the dye FUN® 1. Living yeast cells show red fluorescent vacuolar tubules.
  • Yeast viability dye
  • Red fluorescent tubules only in live cells
  • Diffuse green fluorescence
  • 10 mM solution in DMSO
  • Store at −20 °C, protected from light
  • C24H18ClIN2S
ViaVac™ red/green (also known as FUN®1) is a fluorescent vitality dye for yeast. It forms red fluorescent vacuolar tubules in metabolically active yeast cells.
ViaVac™ dye passively enters cells, giving diffuse, green cytoplasmic staining of both live and dead cells. In metabolically active yeast and fungus cells the dye is actively transported into the vacuole, resulting in red fluorescent staining of tubular intravacuolar structures.
FUN®1 is a registered trademark of Thermo Fisher Scientific.
Customers Who Bought This Also Bought